Learn More
Quickfit™ Two-Neck Round-Bottom Glass Flask
Product Code. 15537860
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Socket Joint:
14/23 (Centre), 14/23 (side)
24/29 (Centre), 14/23 (side)
24/29 (Centre), 19/26 (side)
34/35 (Centre), 19/26 (side)
Unit Size:
Each
Pack of 5
Product Code. 15537860
Brand: Quickfit FR1L/3S/2A
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Two-Neck Round-Bottom Glass Flask has high resistance to thermal shock and chemical attack.
  • With highest-quality Quickfit™ ground glass socket
  • Side neck useful for introducing gas inlet tubes and thermometers etc., into the flask
  • Manufactured from borosilicate glass for highest resistance to thermal shock and chemical attack
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Height (Metric) 180 mm
No. per Case 2
Socket Joint 24/29 (Centre), 19/26 (side)
Bottom Shape Round