Learn More
Quickfit™ Borosilicate Glass Still Head Adapter
Product Code. 11972098
Change view
Click to view available options
Standard Taper Joint:
Cone: 14/23 (Condenser), 14/23 (Flask); Socket: 14/23
Cone: 19/26 (Condenser), 19/26 (Flask); Socket: 14/23
Cone: 19/26 (Condenser), 19/26 (Flask); Socket: 19/26
Cone: 19/26 (Condenser), 24/29 (Flask); Socket: 14/23
Cone: 24/29 (Condenser), 24/29 (Flask); Socket: 14/23
Cone: 29/32 (Condenser), 29/32 (Flask); Socket: 14/23
Cone: 34/35 (Condenser), 34/35 (Flask); Socket: 14/23
Unit Size:
Each
Pack of 5
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint unitSize
11972098 Cone: 14/23 (Condenser), 14/23 (Flask); Socket: 14/23 Each
12068483 Cone: 19/26 (Condenser), 19/26 (Flask); Socket: 14/23 Each
11332115 Cone: 19/26 (Condenser), 19/26 (Flask); Socket: 19/26 Each
12088483 Cone: 24/29 (Condenser), 24/29 (Flask); Socket: 14/23 Each
12078483 Cone: 19/26 (Condenser), 24/29 (Flask); Socket: 14/23 Each
12098483 Cone: 29/32 (Condenser), 29/32 (Flask); Socket: 14/23 Each
15527970 Cone: 34/35 (Condenser), 34/35 (Flask); Socket: 14/23 Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 11972098 Supplier Quickfit Supplier No. SH4/1
Trusted Glassware Deals
Use promo code 28509
View all promotions
Estimated Shipment: 06-10-2026
Add to basket
Add to basket
Connect a flask with a condenser in a distillation assembly. The QuickFit™ still head adapter is designed to accept a thermometer as well.
Connect a flask with a condenser in a distillation assembly. The QuickFit™ still head adapter is designed to accept a thermometer as well.
  • Used in distillation assemblies for connecting flasks to condenser
  • Upper socket designed to accept thermometer either via cone/screw thread adapter or thermometer pocket
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Connecting flask and condenser
Type Still Head Adapter
Material Borosilicate Glass
Standard Taper Joint Cone: 14/23 (Condenser), 14/23 (Flask); Socket: 14/23