Learn More
Quickfit™ Borosilicate Glass Sintered Dip Tube for Dreschel Bottle
Click to view available options
For Use With (Equipment):
For model MF29/23/250 Dreschel bottle
For model MF29/3/125 Dreschel bottle
For model MF29/3/250 Dreschel bottle
For model MF29/3/500 Dreschel bottle
Description:
Stem Length: 132 mm
Stem Length: 165 mm
Stem length: 217 mm
Porosity:
0
1
2
3
Unit Size:
Each
Pack of 5
Description
Disperse gas into bubbles with QuckFit™ sintered borosilicate glass tubes designed to fit a range of QuickFit™ Deschel bottles. Tubes come in a variety of porosity for fine control over bubble size.
- Complies with BS 2461
- Sintered dip tube disperses gas into bubbles for greater efficiency
- Choice of sinter porosities to achieve fine bubble size
Specifications
Specifications
| Certifications/Compliance | BS 2461 |
| Description | Stem Length: 165 mm |
| Material | Borosilicate Glass |
| For Use With (Equipment) | For model MF29/23/250 Dreschel bottle |
| Product Type | Bottle Heads |
| Porosity | 2 |