Learn More
Quickfit™ Borosilicate Glass Single Neck Pear Shaped Flask
Product Code. p-8002651
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
Diameter (Metric) Outer:
23 mm
28 mm
38 mm
48 mm
58 mm
Height (Metric):
67 mm
85 mm
89 mm
100 mm
115 mm
125 mm
135 mm
140 mm
150 mm
Unit Size:
Each
Pack of 5
Product Code. 11912088
Brand: Quickfit FP100/1
Allows distillation down to small quantities.
  • With highest quality Quickfit™ ground glass socket
  • Ideal for semi-micro distillations
  • Pear shaped design allows distillation down to small quantities with minimum hold-up, and reduced losses from thermal decomposition
  • Borosilicate glass for chemical resistance and thermal performance
TRUSTED_SUSTAINABILITY
Specifications
Description Pear
Capacity (Metric) 100 mL
Material Borosilicate Glass
Height (Metric) 135 mm
Diameter (Metric) Outer 58 mm