Learn More
Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask
Product Code. p-8002650
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
150 mL
250 mL
500 mL
1000 mL
2000 mL
3000 mL
5 L
6 L
10 L
Socket Joint:
10/19
14/23
19/26
24/29
29/32
34/35
45/40
Unit Size:
Each
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Product Code. 10459330
Brand: Quickfit FR1L/3S
Use these flasks for a variety of applications. Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask are manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock.
  • Hgh quality Quickfit™ ground glass socket
  • Comprehensive range of 46 flasks from 5mL to 10,000mL
  • Body diameter and wall thickness comply with relevant ISO 1773 and ISO 4797 requirements
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 1773, ISO 4797
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Height (Metric) 180 mm
Neck Style Short
Socket Joint 24/29
Bottom Shape Round