Learn More
Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask
Product Code. p-8002649
Click to view available options
Capacity (Metric):
50 mL
100 mL
150 mL
250 mL
500 mL
1000 mL
2000 mL
4 L
Height (Metric):
85 mm
95 mm
103 mm
105 mm
110 mm
115 mm
130 mm
135 mm
140 mm
145 mm
155 mm
170 mm
175 mm
187 mm
205 mm
215 mm
230 mm
260 mm
Socket Joint:
19/26
24/29
29/32
34/35
Unit Size:
Pack of 10
Pack of 2
Pack of 20
Pack of 5
Product Code. 12498936
Brand: Quickfit FF1L/3US
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
  • Complies with ISO 1773 and ISO 4797
  • With highest quality Quickfit™ ground glass socket
  • Borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • Flat base allows use with hotplates
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 1773, ISO 4797
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Height (Metric) 170 mm
Neck Style Short
Socket Joint 24/29
Bottom Shape Flat