Learn More
Quickfit™ Borosilicate Glass Flange Lid
Product Code. 11301875
Change view
Click to view available options
Description:
0° Side Socket: 14/23;, 5°, 10°, 15° Side Sockets: 19/26, Vertical Center Socket: 19/26
0°, 10° Side Sockets: 19/26, 5° Side Socket: 24/29, Vertical Center Socket: 19/26
0°, 10° and 2 x 5° Side Sockets: 19/26, Vertical Center Socket: 19/26
0°, 2 x 10° Side Sockets: 19/26, Vertical Center Socket: 34/35
0°, 5°, 10° Side Sockets: 14/23, 15° Side Socket: 29/32, Vertical Center Socket: 19/26
0°, 5°, 10° Side Sockets: 19/26, 15° Side Socket: 34/35, Vertical Center Socket:: 19/26
0°, 5°, 10° Side Sockets: 19/26, 15° Side Socket: 34/35, Vertical Center Socket:: 24/29
0°, 5°, 10°, Side Socket: 14/23, 15° Side Socket: 29/32, Vertical Center Socket: 29/32
Vertical Center Socket: 29/32
Vertical Center Socket: 34/35
Closure Size:
75 mm
100 mm
Unit Size:
Each
Pack of 5
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Closure Size Unit Size
11301875 Vertical Center Socket: 29/32 100 mm Each
11362025 0°, 5°, 10° Side Sockets: 19/26, 15° Side Socket: 34/35, Vertical Center Socket:: 19/26 100 mm Each
11342025 0°, 10° and 2 x 5° Side Sockets: 19/26, Vertical Center Socket: 19/26 100 mm Each
11352025 0°, 10° Side Sockets: 19/26, 5° Side Socket: 24/29, Vertical Center Socket: 19/26 100 mm Each
11372025 0°, 5°, 10° Side Sockets: 19/26, 15° Side Socket: 34/35, Vertical Center Socket:: 24/29 100 mm Each
10714537 0°, 5°, 10°, Side Socket: 14/23, 15° Side Socket: 29/32, Vertical Center Socket: 29/32 100 mm Each
11311875 Vertical Center Socket: 34/35 100 mm Each
10064184 0° Side Socket: 14/23;, 5°, 10°, 15° Side Sockets: 19/26, Vertical Center Socket: 19/26 75 mm Each
11321875 Vertical Center Socket: 34/35 75 mm Each
15557920 0°, 5°, 10° Side Sockets: 14/23, 15° Side Socket: 29/32, Vertical Center Socket: 19/26 100 mm Pack of 5
15567920 0°, 2 x 10° Side Sockets: 19/26, Vertical Center Socket: 34/35 100 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 11301875 Supplier Quickfit Supplier No. DA4/100
Estimated Shipment: 17-12-2026
Constructed of Borosilicate Glass. Quickfit™ Borosilicate Glass Flange Lid includes precision ground flat-flange joints. Fits Quickfit™ wide neck reaction flasks and culture vessels. Each lid with one or more sockets to accept other Quickfit™ components.
Constructed of Borosilicate Glass. Quickfit™ Borosilicate Glass Flange Lid is comprised of precision ground flat-flange joints. Fits Quickfit™ wide neck reaction flasks and culture vessels. Each lid includes one or more sockets to accept other Quickfit™ components.
  • With precision ground flat-flange joints
  • Fits Quickfit™ wide neck reaction flasks and culture vessels
  • Each lid with one or more sockets to accept other Quickfit™ components
Specifications
Description Vertical Center Socket: 29/32
Material Borosilicate Glass
For Use With (Equipment) Wide neck flasks
Includes Sockets for use with other Quickfit™ components
Product Type Flange lid
Closure Size 100 mm