Learn More
Quickfit™ Borosilicate Glass Drying Angled Tube
Click to view available options
Standard Taper Joint No.:
14/23
19/26
24/29
29/32
Unit Size:
Each
Pack of 5
Description
- For use with a desiccant such as calcium chloride
- Ground glass cone to fit Quickfit™ apparatus
Specifications
Specifications
| For Use With (Equipment) | Quickfit™ apparatus |
| Material | Borosilicate Glass |
| Product Type | Tube |
| Standard Taper Joint No. | 19/26 |