Learn More
Pyrex™ Quickfit™ Pear Shaped Distillation Flask
Product Code. 15587920
Change view
Click to view available options
Capacity (Metric):
50 mL
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15587920 50 mL Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15587920 Supplier Quickfit Supplier No. MD2/50
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Designed for micro and semi-micro distillations
  • Vertical neck with Quickfit socket for introducing a thermometer
  • Angled cone for connecting to condenser
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 50 mL
Material Borosilicate Glass
For Use With (Application) Micro and semi-micro distillations
Height (Metric) 147 mm
Neck Style Vertical
Shape Pear
Socket Joint 14/23