Learn More
Pyrex™ Quickfit™ Borosilicate Glass Cold Trap Head
Product Code. 10536465
Change view
Click to view available options
Standard Taper Joint:
34/35, 19/26
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint Unit Size
10536465 34/35, 19/26 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10536465 Supplier Pyrex™ Supplier No. MF28/52
  • ONLINE EXCLUSIVE PRICE
    135.45€
    112.04€ / Pack of 1
    Save 23.41€
    17% Off
Estimated Shipment: 18-01-2027
Freezes volatile materials and vapors onto the cold surface. Pyrex™ Quickfit™ Borosilicate Glass Cold Trap Head prevents materials from damaging vacuum pumps.
  • Designed for use in vacuum systems
  • Prevents vapours from volatile materials from damaging pumps etc
  • Easy to use: - immerse trap into Dewar vessel containing refrigerant e.g., dry ice - volatile materials passing through system are frozen onto the cold surface
Specifications
Description Cold trap head
Material Borosilicate Glass
For Use With (Equipment) VAcuum systems
Standard Taper Joint 34/35, 19/26