Learn More
Quickfit™ Borosilicate Glass Büchner Vacuum Filtration Funnels with Glass Sintered Disc
Useful where conventional filter papers would be destroyed by acids and alkalis
Brand: Quickfit VB60/08M
This item is not returnable.
View return policy
Description
- Manufactured from borosilicate glass for highest resistance to chemical attack
- With Quickfit ground glass cone to allow direct filtration into Quickfit Büchner flasks