Learn More
3M™ Quick Latch Head Harness Assembly
Product Code. 15769067 Shop All 3M Products
Change view
Click to view available options
Color: Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15769067 Color: Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15769067 Supplier 3M™ Supplier No. 7100020701
  • ONLINE EXCLUSIVE PRICE
    429.00€
    292.83€ / Pack of 20
    Save 136.17€
    31% Off
Estimated Shipment: 29-01-2027
3M™ Quick Latch Head Harness Assembly is a replacement part for use with 3M™ Reusable Half Face Mask 6500 Series.
  • Replacement Quick Latch Head Harness Assembly
  • For use with the 3M™ Half Face Mask 6500 Series
  • Replace worn out or damaged parts
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Product Type Head Harness
For Use With (Equipment) 3M™ Reusable Half Face Mask 6500 Series