Learn More
Thermo Scientific Chemicals Quartz tubing, fused, 12mm (0.472in) OD, 8mm (0.315in) ID
Product Code. 15411797
Click to view available options
Diameter (Metric) Outer:
6.35 mm
12 mm
12.7 mm
19 mm
25 mm
Overall Length (Metric):
30.4 cm
122 cm
Unit Size:
120cm
30cm
Product Code. 15411797
Brand: Thermo Scientific Chemicals 042272.VB
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Manufactured by fusing naturally occurring crystalline silica
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 8 mm
Material Fused Quartz
Diameter (English) Outer 0.472 in.
Diameter (Metric) Outer 12 mm
Overall Length (Metric) 122 cm
Product Type Fused Quartz Tubing