Learn More
Hellma™ Quartz Suprasil Ultra-Micro Absorption Cells
Specifications
Specifications
| Type | Ultra-micro Absorption Cell |
| Capacity (Metric) | 100 μL |
| Material | Quartz Suprasil |
| Quantity | 2/Pack |
| Path Length | 3 x 3 mm |
| Height (Metric) | 45 mm |
| Width (Metric) | 12.5 mm |