Learn More
Hellma™ Quartz Suprasil Sealable Cells
Click to view available options
Capacity (Metric):
3.5 mL
Unit Size:
Each
Specifications
Specifications
| Type | Sealable Cell |
| Capacity (Metric) | 3.5 mL |
| Material | Quartz Suprasil |
| Closure Material | PTFE |
| Width (Metric) Interior | 13mm |
| Quantity | 1Each |
| Path Length | 10 mm |
| Wavelength Range | 200 to 2500 nm |
| Height (Metric) | 56 mm |
| Width (Metric) | 12.5 mm |