Learn More
Helios Italquartz Quartz Evaporating Dishes
Product Code. p-8000258
Click to view available options
Capacity (Metric):
45 cc
85 mL
Unit Size:
Each
Product Code. 11715694
Brand: Helios Italquartz 61F00141
Excellent for producing concentrated solutions or solid precipitates of a dissolved substance. Helios Italquartz™ Quartz Evaporating Dishes are resistant to thermal shock up to 1000°C.
  • Resistant to corrosion
  • Chemically inert to acids (except HF)
  • Without spout
Specifications
Capacity (Metric) 45 cc
Material Quartz
Shape Round
Type Evaporating Dish
Height (Metric) 28 mm
Diameter (Metric) 56 mm