Learn More
Bel-Art™ SP Scienceware™ Wilmad™ X-Band Capillary EPR Tubes for Aqueous Sample
Product Code. 15348608 Shop All SP Scienceware Products
Click to view available options
Length (Metric):
100 mm
Unit Size:
Pack of 25
Product Code. 15348608
Brand: Bel-Art™ 660000040
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Close up the end with a hot flame or with beeswax. Bel Art SP Scienceware™ Wilmad™ X-Band Capillary EPR Tube for Aqueous Sample are often used in EPR applications that require tuning of aqueous samples at high dB or where sample size is limited.
  • Open on both ends
  • Easy to seal with a hot flame or beeswax
Specifications
Length (Metric) 100 mm
Diameter (Metric) Outer 1.2 mm
Diameter (Metric) Inner 1.0 mm
Quantity 25 Pack
Model Q-1.0X1.2