Learn More
Thermo Scientific Chemicals Quartz crucible, fused; Ht(mm), 57; Top OD(mm), 43; Base OD(mm), 22; Volume(ml), 30
Product Code. 15473895
Change view
Click to view available options
Capacity (Metric):
30 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15473895 30 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15473895 Supplier Thermo Scientific Chemicals Supplier No. 043498.KG
 Restricted Item
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Manufactured by fusing naturally occurring crystalline silica
  • High chemical purity
  • High corrosion resistance
  • High melting point, extreme hardness
  • Low coefficient of thermal expansion
  • Excellent electrical insulation qualities, and optical transmission from ultra-violet to infra-red
TRUSTED_SUSTAINABILITY
Specifications
Material Quartz
Capacity (Metric) 30 mL
Diameter (Metric) Outer Top: 43mm; Bottom: 22mm
Height (Metric) 57mm
Product Type Fused quartz crucible