missing translation for 'onlineSavingsMsg'
Learn More

Motic™ Quarter Lambda Wave Retarder Plate

Product Code. 17129378
Click to view available options
For Use With (Equipment):
Motic TEC POL Microscopes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17129378

Brand: Motic™ 1101006700165

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Plate retards light by one quarter of a wave for TEC POL Microscopes.

Retards light by one quarter of a wave. For use with the following Motic™ Microscopes:

  • Panthera TEC POL Binocular
  • Panthera TEC POL Trinocular
  • Panthera TEC POL Epi Binocular
  • Panthera TEC POL Epi Trinocular
  • Panthera TEC POL Digital
  • Panthera TEC POL Epi Digital
  • Panthera TEC POL (i)

Specifications

Type Waveplate
Color Black
For Use With (Application) Retarding Microscope Light
For Use With (Equipment) Motic TEC POL Microscopes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.