Learn More
Applied Biosystems™ QuantStudio™ 12K Flex OpenArray™ Gene Expression Starter Kit
A variety of starter kits are available to introduce you to the various applications of the QuantStudio™12K Flex OpenArray™ sample block.
Brand: Applied Biosystems™ 4469604
Description
Each starter kit contains 2 arrays, plus sufficient reagents and accessories to perform two experiments. The Gene Expression starter kit contains 2 gene expression endogenous control plates. The Gene Expression kit also contains cDNA controls to serve as samples for your experiments. Note that the QuantStudio™12K Flex Software (included with the QuantStudio™ 12K Flex System) offers example experiment files for each application.
- PCR Method: qPCR
- Built-In Hot Start Primer-Probe Compatability: TaqMan™
Specifications
| • 2X TaqMan™ OpenArray™ Real-Time PCR Master Mix, 1.5 mL • TaqMan™ OpenArray™ Human Endogenous Control Panels (2 plates) • Human cDNA controls (brain, liver, lung, and placenta) • QuantStudio™ 12K Flex OpenArray™ Accessories Starter Kit |
|
| qPCR | |
| QuantStudio™ 12k Flex | |
| 12K Flex Gene Expression Starter Kit | |
| cDNA |
| Primer-Probe | |
| Standard | |
| OpenArray™, QuantStudio™ | |
| Built-In Hot Start |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.