Learn More
Macherey-Nagel™ Quantofix™ Nitrite Test Strip Kits
Description
Allows for the quick and easy determination of nitrite in solutions. Macherey Nagel™ Quantofix™ Nitrite Test Strip Kits are semi-quantitative, come pre-calibrated and contain all necessary equipment and reagents.
- Ready to use
- Rapid: provides reliable results within one minute
- Excellent for checking cooling lubricants and aquaculture/fish farming
- Precise: colour charts adjusted and check using certified standard solutions
- Shelf life: 2.5 years, store below 30°C
Specifications
Specifications
| Color Indicator | Yellow to Red |
| Format | Strip |
| For Use With (Application) | Determination of Nitrite |
| Graduations | 0, 1, 5 ,10, 20, 40, 80mg/L NO 2 - |
| Quantity | 100 Pack |
| Storage Requirements | 30°C |
| Test Time | 1 min. |
| Type | Semi-qualitative Test Strip |