missing translation for 'onlineSavingsMsg'
Få mere at vide

Invitrogen™ QuantiGene™ Sample Processing Kit, tissues

Artikelnummer. 31941713
Skift visning
Klik for at se tilgængelige muligheder
Quantity:
100 Samples
Pakningsstørrelse:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity Pakningsstørrelse
31941713 100 Samples Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 31941713 Leverandør Invitrogen™ Leverandørnr. QS0106
 Restricted Item
Estimated Shipment: 21-10-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

The QuantiGene Sample Processing Kit for tissues contains reagents and instructions for the preparation of tissue homogenates from fresh or frozen animal or plant tissues for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.

The QuantiGene Sample Processing Kit for tissues contains reagents and instructions for the preparation of tissue homogenates from fresh or frozen animal or plant tissues for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.

  • Easy to follow instructions
  • Preserves the integrity of the RNA/DNA without degradation
  • Choose the method that matches your tissue sample

What you get

The QuantiGene Sample Processing Kit for tissues contains our proprietary homogenizing solution, proteinase K, and detailed instructions for preparing homogenized tissue from a variety of tissue sources.

TRUSTED_SUSTAINABILITY

Tekniske data

Content And Storage • 75 mL Homogenizing Solution, store at 15°C to 30°C
• 360 μL Proteinase K (50 μg/μL), store at -20°C

Format Kit
Sample Type Tissue
Product Line QuantiGene
Product Type Sample Processing Kit
Quantity 100 Samples
Shipping Condition Blue Ice
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.