missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QuantiGene™ Sample Processing Kit, FFPE samples

Product Code. 31942263
Change view
Click to view available options
Quantity:
100 Samples
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31942263 100 Samples Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31942263 Supplier Invitrogen™ Supplier No. QS0109
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The QuantiGene Sample Processing Kit for formalin-fixed paraffin-embedded (FFPE) samples contains reagents and instructions for the preparation of tissue homogenates from FFPE tissue samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets.

The QuantiGene Sample Processing Kit for formalin-fixed paraffin-embedded (FFPE) samples contains reagents and instructions for the preparation of tissue homogenates from FFPE tissue samples for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA or DNA targets. H&E stained slides are compatible with QuantiGene assays.

  • Easy to follow instructions
  • Preserves the integrity of the RNA/DNA without degradation
  • Choose the method that matches your FFPE tissue sample

What you get

The QuantiGene Sample Processing Kit for FFPE samples contains our proprietary homogenizing solution, proteinase K, and detailed instructions for preparing tissue homogenates from FFPE tissue sections.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 75 mL Homogenizing Solution, store at 15°C to 30°C
• 360 μL Proteinase K (50 μg/μL), store at -20°C

Format Kit
Sample Type FFPE
Product Line QuantiGene
Product Type Sample Processing Kit
Quantity 100 Samples
Shipping Condition Blue Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.