missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QuantiGene™ Sample Processing Kit, cultured cells

Product Code. 31942695
Change view
Click to view available options
Quantity:
5 x 10 Plates
50 Plates
Unit Size:
Each
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31942695 5 x 10 Plates Each
31942259 50 Plates Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 31942695 Supplier Invitrogen™ Supplier No. QS0102
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This QuantiGene Sample Processing Kit for cultured cells contains reagents and instructions for the preparation of cell lysates for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA targets and QuantiGene Plex DNA assays for DNA targets.

This QuantiGene Sample Processing Kit for cultured cells contains reagents and instructions for the preparation of cell lysates for use in QuantiGene Singleplex and QuantiGene Plex assays for RNA targets and QuantiGene Plex DNA assays for DNA targets.

  • Easy to follow instructions
  • Preserves the integrity of the RNA/DNA without degradation
  • Choose the method that matches your cultured cell sample

What you get
The QuantiGene Sample Processing Kit for cultured cells contains our proprietary lysis mixture, proteinase K, and detailed instructions for preparing cell lysates from cultured cells.

Success in cell lysis
Key factors that influence the complete lysis of the cells include:

  • The lysis mixture
  • Proteinase K
  • Vortexing or pipetting of the sample
  • Storage of the lysates at -80°C
  • Incubation at 50°C

The protocols provided with the QuantiGene Sample Processing Kit for cultured cells use a combination of these factors to ensure complete lysis of the samples. In addition, the lysis mixture provides the proper ionic conditions for the overnight hybridization in the QuantiGene assays.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 5 x 75 mL Lysis Mixture, store at 15°C to 30°C
• 5 x 750 μL Proteinase K (50 μg/μL), store at -20°C

Format Kit
Sample Type Cell Cultures
Product Line QuantiGene
Product Type Sample Processing Kit
Quantity 5 x 10 Plates
Shipping Condition Blue Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.