Learn More
Thermo Scientific™ Quanti-Cult™ Escherichia coli ATCC™ 25922™
Product Code. 12657699
Change view
Click to view available options
Description:
Escherichia coli ATCC™ 25922™
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description unitSize
12657699 Escherichia coli ATCC™ 25922™ Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12657699 Supplier Thermo Scientific™ Supplier No. R4737050
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
Obtain fast and accurate identification of microbiological organisms with real biological organisms growing on fresh enriched microbiological culture media.
Thermo Scientific™ Quanti-Cult™ Escherichia coli ATCC™ 25922™ is supplied as a film of preserved microorganisms on the inside of the cap on a plastic vial, ready for rehydration and use in quality control procedures.
  • No serial dilution required
  • Delivers <100 colony forming units (CFUs) in a single 0.3mL inoculum
Ordering Alerts:
1 Test/reconstituted vial, 10 Tests/kit
Includes:
10 preserved organism vials and 10 rehydration vials.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 25922TM*
Description Escherichia coli ATCC™ 25922™
Format Quanti-Cult
Product Type Bactidisk
Target Organism Class Gram-negative Rods
Test Type Quantitative QC
Packaging Type 10 Test/Kit
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.