Learn More
Socorex™ Qualitix™ Sterilization Paper Tray
Product Code. 17296800
Change view
Click to view available options
Length (English):
10.5 in.
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) unitSize
17296800 10.5 in. Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17296800 Supplier Socorex™ Supplier No. 3220.1526
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Advantageously replacing stainless steel trays, Qualitix™ Sterilization paper tray are fully autoclavable by either EO, Steam or Gamma Ray.
  • Rigid medical PE coated paper material
  • Advantageously replacing stainless steel trays
  • One size fits most devices
  • Suitable for EO, steam and gamma sterilization
  • Fit for use as such, or in combination with Qualitix™ sterilization pouches
  • Conformity with ISO 14971, ISO 11135, ISO 17665-06, CE standard
Specifications
Autoclavable Autoclavable
Certifications/Compliance ISO 14971, ISO 11135, ISO 17665-06, CE standard
Color White
Disposable Single-use
For Use With (Application) Sterilization
For Use With (Equipment) Autoclave or EO or Gamma Rays
Includes Paper tray only
Material Polyethylene-coated Medical Paper
Sterility Non-sterile
No. per Case 500
No. per Pack 50
Dimensions (L x W x H) 150 x 265 x 15 mm
Length (English) 10.5 in.
Width (English) 6 in.
Height (English) 0.6 in.
Height (Metric) 15 mm
Product Type Sterilization tray
Length (Metric) 265 mm
Width (Metric) 150 mm