Learn More
Munktell Filtrak™ Qualitative Ashless Papers; Circle
Click to view available options
Diameter (Metric):
150 mm
Particle Retention:
>20 μm
5 to 6 μm
Unit Size:
Pack of 100
Description
- Very fast filtering, open fiber matrix
- Fast filtering, open to semi-open fiber matrix
- Medium to fast filtering, semi-open to open fiber matrix
- Medium to slow filtering, medium-tight fiber matrix
- Very slow filtration, very tight fiber matrix
Specifications
Specifications
| Material | Cellulose |
| Particle Retention | 5 to 6 μm |
| Filtration Speed | 29s/10mL |
| For Use With (Application) | Fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulfate |
| Grade | V1F |
| Diameter (Metric) | 150 mm |