Learn More
Hellma™ Suprasil™ Quartz Micro Absorption Cell Cuvettes
Click to view available options
Capacity (Metric):
14,000 μL
17,500 μL
35,000 μL
7000 μL
Unit Size:
Each
Pack of 2
Description
- Very chemically resistant
- Maintains shape, even at high temperatures up to approximately 1000°C
- 80% transmission from 320 to 2500nm
Specifications
Specifications
| Type | Macro Absorption Cell |
| Capacity (Metric) | 7000 μL |
| Material | Quartz Suprasil |
| Width (Metric) Interior | 9.5mm |
| Path Length | 20 mm |
| Wavelength Range | 200 to 2500 nm |
| Height (Metric) | 45 mm |
| Width (Metric) | 12.5 mm |