Learn More
STEMCELL Technologies qPCR Master Mix Kit
For probe-based assays and arrays
Brand: STEMCELL Technologies 07516
Additional Details : Weight : 0.30000kg
Description
- qPCR Master Mix is a 2X concentrated solution optimized for TaqMan™ probe-based, real-time quantitative polymerase chain reaction (qPCR)
- It is intended for use in combination with gene-specific primers (to amplify target cDNA) and fluorogenic-labeled probes that use the 5' nuclease activity of Taq DNA polymerase to produce a fluorescent signal
- The rate of accumulation of the fluorescent signal is used to quantify the cDNA, and thereby to determine the amount of mRNA present in the original sample
- qPCR Master Mix includes a Hot Start DNA polymerase, dNTPs, MgCl2, enhancers, and stabilizers
- The qPCR Master Mix Kit also includes ROX Reference Dye as a separate component, making it compatible with both reference dye-dependent and -independent qPCR systems
- Efficient, sensitive, and reproducible
- Optimal performance when using standard or fast cycling conditions
- Ideal for high-throughput applications and overnight experiments
- Compatible with various real-time qPCR instruments
Specifications
Adipocytes; Chondrocytes; Endoderm, PSC-Derived; Hepatic Cells; Intestinal Cells; Mesenchymal Stem and Progenitor Cells; Osteoblasts; Pancreatic Cells; Pluripotent Stem Cells | |
1 mL Kit | |
Real Time PCR (qPCR) |