missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ qPCR& Go Probe Hi-ROX Kit
Fast and sensitive qPCR Probe Master Mix. For most ABI platforms
Brand: MP Biomedicals™ 11EBI04200
Additional Details : Weight : 0.00500kg
Description
- The qPCR& Go Probe Hi-ROX Kit has been developed for fast, highly reproducible real-time PCR and has been validated on commonly used real-time PCR instruments.
- The kit has been formulated for use with probe-detection technology, including TaqMan™, Scorpions™ and molecular beacon probes.
- A combination of the latest advances in buffer chemistry and PCR enhancers, together with a hot-start DNA polymerase, ensures that the qPCR& Go Probe Kit delivers fast, highly-specific and ultra-sensitive real-time PCR.
- qPCR& Go Probe is provided as a 2x mastermix containing all the components necessary for real-time PCR, including dNTPs, stabilizers and enhancers.
Specifications
dNTPs, stabilizers and enhancers. Store at -20°C | |
qPCR Kit | |
Real Time PCR (qPCR) | |
500 |
Fast and sensitive qPCR Probe Master Mix. For most ABI platforms. | |
4 x 5 mL | |
2x |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.