missing translation for 'onlineSavingsMsg'
Learn More

Pyruvaldehyde (35% w/w aqueous), TRC

Product Code. 30010938
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010938 10 g 10g
30003437 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010938 Supplier Toronto Research Chemicals Supplier No. TRCP99887010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-98-8
Molecular Formula C3 H4 O2
Molecular Weight (g/mol) 72.06
Synonym Propanal, 2-oxo-, Pyruvaldehyde (8CI), 2-Oxopropanal, 2-Ketopropionaldehyde, 2-Oxopropionaldehyde, Acetylformaldehyde, Acetylformyl, Methylglyoxal, NSC 626580, NSC 79019, Pyroracemic aldehyde, Pyruvic aldehyde, α-Ketopropionaldehyde, Propanal, 2-oxo-, Pyruvaldehyde (8CI), 2-Oxopropanal, 2-Ketopropionaldehyde, 2-Oxopropionaldehyde, Acetylformaldehyde, Acetylformyl, Methylglyoxal, NSC 626580, NSC 79019, Pyroracemic aldehyde, Pyruvic aldehyde, α-Ketopropionaldehyde
IUPAC Name 2-oxopropanal
SMILES CC(=O)C=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C3H4O2/c1-3(5)2-4/h2H,1H3
Formula Weight 72.0211
Chemical Name or Material Pyruvaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.