Learn More
Pyrrole-3-carboxaldehyde, 97%
Product Code. 15497537
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15497537 1 g 1g
15407547 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15497537 Supplier Thermo Scientific Alfa Aesar Supplier No. H35559.03
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
CAS: 7126-39-8 | C5H5NO | 95.10 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7126-39-8
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.10
MDL Number MFCD03426733
InChI Key CHNYVNOFAWYUEG-UHFFFAOYSA-N
Synonym pyrrole-3-carboxaldehyde, 1h-pyrrole-3-carboxaldehyde, pyrrole-3-carbaldehyde, 3-formylpyrrole, 3-formyl-1h-pyrrole, 3-formyl-pyrrole, pyrrol-3-aldehyde, pyrrole-3-aldehyde, pubchem18051, acmc-209oht
PubChem CID 640308
IUPAC Name 1H-pyrrole-3-carbaldehyde
SMILES O=CC1=CNC=C1
Specifications
Melting Point 65°C to 68°C
Quantity 1 g
Sensitivity Air sensitive
Formula Weight 95.1
Percent Purity 97%
Chemical Name or Material Pyrrole-3-carboxaldehyde