Learn More
Tocris Bioscience™ Pyrintegrin
Description
Pyrintegrin enhances survival of human embryonic stem cells (hESCs) following enzymatic dissociation. Thought to enhance cell-ECM adhesion and activate integrin signaling. Pyrintegrin increases adhesion of hESCs to matrigel- and laminin-coated plates, but not to gelatin-coated plates. Also podocyte-protective agent and prevents LPS-induced proteinuria in mice.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | Pyrintegrin |
| Purity | >98% |
| Molecular Weight (g/mol) | 451.54 |
| Inhibitors | Pyrintegrin |