missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Pyrintegrin

Product Code. 17150132
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17150132 1 mg 1mg
17160132 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17150132 Supplier STEMCELL Technologies Supplier No. 72842

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Integrin and tyrosine kinase activator; Activates Integrin, FGFR, IGFR, and HER2

  • Pyrintegrin is a 2,4-disubstituted pyrimidine that induces the activation of β1 integrin and multiple growth factor receptors, including FGFR1, IGFR1, EGFR1, and HER2 (Xu et al.)
  • Enhances integrin-dependent attachment and survival of human embryonic stem (ES) cells following trypsin-mediated single-cell dissociation (Xu et al.)

Specifications

CAS 1228445-38-2
Quantity 1 mg
Molecular Formula C23H25N5O3S
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.