missing translation for 'onlineSavingsMsg'
Learn More

Pyridinium fluorochromate, 98%

Product Code. 11344186
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11344186

Brand: Thermo Scientific Alfa Aesar L19295.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 83042-08-4 | C5H6CrFNO3

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83042-08-4
Molecular Formula C5H6CrFNO3
MDL Number MFCD00151617
Synonym pyridin-1-ium; trioxochromium; hydrofluoride, chromium trioxide pyridium hydrofluoride, pyridin-1-ium; tris oxidanylidene chromium; hydrofluoride

Specifications

Melting Point 165°C (decomposition)
Quantity 25 g
UN Number UN1479
Sensitivity Moisture sensitive
Formula Weight 199.1
Percent Purity 98%
Chemical Name or Material Pyridinium fluorochromate
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1A
  • Oxidising solid Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H317-May cause an allergic skin reaction.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.