missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyridine, for DNA Synthesis And Protein Sequencing, Anhydrous, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 L
Packaging:
Plastic Coated Glass bottle
Unit Size:
1 L
Specifications
Specifications
| CAS | 110-86-1 |
| Melting Point | -42°C |
| Color | Colorless |
| pH | 8.5 |
| Boiling Point | 115°C |
| Packaging | Plastic Coated Glass bottle |
| Molecular Formula | C5H5N |
| MDL Number | 11732 |
| Quantity | 1 L |
| Synonym | azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction