Learn More
Pyrex™ Separating Funnel with Long Delivery Stem and PTFE Key
Product Code. 12519165
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
2000 mL
Stopcock:
(Bore Size) 1.5mm
(Bore Size) 2.5mm
(Bore Size) 4mm
Joint Number:
14/23
19/26
24/29
29/32
Unit Size:
Each
Product Code. 12519165
Brand: Pyrex 443042N
Supplied with hollow glass stopper and interchangeable PTFE key for grease-free performance. Pyrex™ Separating Funnel with Long Delivery Stem and PTFE Key provides good control of the liquid/liquid interface.
  • Gilson style with long delivery stem for good control of the liquid/liquid interface
  • With interchangeable PTFE key for grease-free performance
  • Supplied with hollow glass stopper
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 125 mL
Material Borosilicate Glass
Includes Hollow Glass stopper, PTFE key
Joint Number 19/26
Stopcock (Bore Size) 1.5mm
Product Type Separatory Funnel