Learn More
Pyrex™ Scoop with Hollow Stem
Product Code. p-8002614
Click to view available options
Capacity (Metric):
3 mL
6 mL
10 mL
Height (Metric):
70 mm
85 mm
100 mm
Unit Size:
Pack of 10
Product Code. 12651992
Brand: Pyrex 2115/06M
Aids the introduction of powder into narrow neck vessels. Pyrex™ Scoop with Hollow Stem sits flat to ensure stability on the balance pan. Available in 3 capacities.
  • With hollow stem to aid introduction of the powder into narrow neck vessels e.g., volumetric flasks
  • The flat base ensures stability on the balance pan
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 10 mL
Material Borosilicate Glass
Height (Metric) 100 mm
Product Type Weighing scoop