Learn More
Pyrex™ Plain Arms, Straight Bore Stopcock
Product Code. 11352115
Click to view available options
Diameter (Metric) Side Arm:
8 mm
9 mm
12.5 mm
Unit Size:
Each
Product Code. 11352115
Brand: Pyrex SSD102
Only null left
Add to basket
Add to basket
Externally operated valve to regulate the flow of a liquid.
  • Glass stopcocks which meet specifications of DIN 12541
  • Manufactured from borosilicate glass with interchangeable key
  • Leakage rate meets the requirements of DIN 12540 i.e. 0.1mbar l/s-1
  • Supplied complete with key retaining device
TRUSTED_SUSTAINABILITY
Specifications
Type Straight Bore Stopcock
Plug Bore 1.5 mm
Diameter (Metric) 12.5 mm
Material Borosilicate Glass
Diameter (Metric) Side Arm 12.5 mm