Learn More
Corning™ PYREX™ Disposable Individually Wrapped Glass Serological Pipets, Sterile
Shop All Corning ProductsDescription
These pipets, calibrated 'To Deliver' with blow-out, offer long, slender, tapered tips to make pipetting go faster and easier. All pipets have negative graduations and fire-polished tips for burr-free, even flow rates. Easy-open Steri-ViewTM wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size. Colors conform to ISO standards. Reference: ASTM E-714.
Do not pipet by mouth. We suggest using a mechanical pipetting device.
Order Info
Individually packaged in easy-open Steri-View™ wrappers or multipacked in plastic bags.
Specifications
Specifications
| Certifications/Compliance | ASTM E714 |
| Material | Glass |
| Graduated | Yes |
| Graduations | 0.01 mL |
| Negative Graduations | 0.2 mL |
| Tip Style | Round |
| Color | Yellow |
| Volume (Metric) | 1 mL |
| Type | Serological Pipet |
| Disposable | Single-use |