Learn More
Corning™ PYREX™ Disposable Individually Wrapped Glass Serological Pipets, Sterile
Product Code. 10494343 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
Graduations:
0.01 mL
0.1 mL
Color:
Blue
Green
Orange
Yellow
Negative Graduations:
0.2 mL
1 mL
2 mL
Unit Size:
Pack of 100
Pack of 120
Pack of 180
Pack of 600
Pack of 720
Pack of 800
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Graduations Color Negative Graduations unitSize
10494343 5 mL 0.1 mL Blue 1 mL Pack of 720
13003093 2 mL 0.01 mL Green 0.2 mL Pack of 180
10776133 2 mL 0.01 mL Green 0.2 mL Pack of 720
11313845 10 mL 0.1 mL Orange 2 mL Pack of 100
10785573 10 mL 0.1 mL Orange 2 mL Pack of 600
11303845 5 mL 0.1 mL Blue 1 mL Pack of 120
10526493 1 mL 0.01 mL Yellow 0.2 mL Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10494343 Supplier Corning™ Supplier No. 70775N
Only null left
Add to basket
Add to basket
Calibrated to deliver (TD) with blow-out. Corning™ Pyrex™ Disposable Individually Wrapped Glass Serological Pipets offer long, slender, tapered tips to make pipetting go faster and easier.
These pipets, calibrated 'To Deliver' with blow-out, offer long, slender, tapered tips to make ­pipetting go faster and easier. All pipets have negative graduations and fire-polished tips for ­burr-free, even flow rates. Easy-open Steri-ViewTM wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size. Colors conform to ISO standards. Reference: ASTM E-714.
Do not pipet by mouth. We suggest using a mechanical pipetting device.
Order Info
Individually packaged in easy-open Steri-View™ wrappers or multipacked in plastic bags.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ASTM E714
Material Glass
Graduated Yes
Graduations 0.1 mL
Negative Graduations 1 mL
Tip Style Round
Volume (English) 0.16 oz.
Color Blue
Volume (Metric) 5 mL
Type Serological Pipet