Learn More
Pyrex™ Dessicator Plates
Click to view available options
Description:
Plate Size: 150 mm
Plate Size: 200mm
Unit Size:
100mL
Each
Description
For use with Pyrex vacuum and non-vacuum desiccators
- For use with Pyrex vacuum and non-vacuum desiccators
Specifications
Specifications
| Description | Plate Size: 150 mm |
| Material | Metal |
| For Use With (Equipment) | Pyrex vacuum and non-vacuum desiccator |