Learn More
Pyrex™ Graduated Dropping Funnel with Glass Key
Product Code. 11313684
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
11313684 250 mL Each
11333684 1000 mL Each
11323684 500 mL Each
11303684 100 mL Each
12433481 50 mL Each
5 options
Product Code. 11313684
Brand: Pyrex 3590/04
Only null left
Add to basket
Add to basket
Used to transfer fluids.
  • Complies with ISO 4800 type 4
  • Manufactured from Pyrex borosilicate glass
  • Double graduated scale showing approximate volume contained/dispensed
  • Graduations in white enamel
  • With interchangeable glass key
  • Supplied with glass stopper
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Graduated Yes
Includes Glass stopper and key
Graduations 5mL
Product Type Dropping Funnel