Learn More
Pyrex™ Borosilicate Glass Wide Neck Flat Bottom Boiling Flask
Product Code. 12423431
Click to view available options
Capacity (Metric):
50 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Pack of 10
Pack of 30
Product Code. 12423431
Brand: Pyrex 1080/14D
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Manufactured from Pyrex™ glass for high resistance to thermal shock
  • Complies with ISO 24450
  • With wide neck to facilitate easy addition of solid samples etc
  • Flat base for standing on benches and hotplates etc
  • Manufactured from Pyrex™ glass for high resistance to thermal shock
  • Excellent chemical durability for contamination free performance
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 24450
Capacity (Metric) 2000 mL
Material Borosilicate Glass
Height (Metric) 230 mm
Neck Style Wide
Diameter (Metric) Outer 166 mm