Learn More
Pyrex™ Borosilicate Glass Thimble
Product Code. 15517730
Change view
Click to view available options
Diameter (Metric) Outer:
26 mm
36 mm
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Unit Size
15517730 36 mm Pack of 10
15507730 26 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15517730 Supplier Pyrex Supplier No. 3845/02M
  • ONLINE EXCLUSIVE PRICE
    339.15€
    284.46€ / Pack of 10
    Save 54.69€
    16% Off
Estimated Shipment: 25-01-2027
Pyrex™ Borosilicate Glass Thimble is highly resistant to chemical attack.
  • Borosilicate glass with sintered glass disc to support solid sample
  • Useful for chemical applications where weighing extracted material is required
Specifications
Diameter (Metric) Outer 36 mm
Material Borosilicate Glass
For Use With (Application) Useful for chemical applications where it is required to weigh the extracted material
Grade Porosity 1
Includes Sintered Glass disc
Length (Metric) 95 mm
Product Type Filter Thimble