Learn More
Pyrex™ Borosilicate Glass Plain Neck Kjeldahl Flask
Product Code. 15547680
Change view
Click to view available options
Capacity (Metric):
100 mL
300 mL
500 mL
800 mL
Diameter (Metric) Outer:
101 mm
115 mm
81 mm
Bulb: 58 mm
Unit Size:
Pack of 10
Pack of 20
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Outer Unit Size
15547680 300 mL 81 mm Pack of 20
11313624 800 mL 115 mm Pack of 10
11303624 500 mL 101 mm Pack of 10
11383614 100 mL Bulb: 58 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15547680 Supplier Pyrex Supplier No. 1200/10D
  • ONLINE EXCLUSIVE PRICE
    621.60€
    516.75€ / Pack of 20
    Save 104.85€
    16% Off
Estimated Shipment: 02-11-2026
Suitable for the determination of nitrogen
  • Withstands high temperatures
  • Resistant to chemical corrosion
Specifications
Diameter (Metric) Inner Neck 24 mm
Diameter (Metric) Outer Neck 28 mm
Capacity (Metric) 300 mL
Material Borosilicate Glass
Height (Metric) 300 mm
Neck Style Plain
Diameter (Metric) Outer 81 mm