Learn More
Pyrex™ Borosilicate Glass Plain Neck Kjeldahl Flask
Product Code. 11303624
Click to view available options
Capacity (Metric):
100 mL
300 mL
500 mL
800 mL
Diameter (Metric) Outer:
101 mm
115 mm
81 mm
Bulb: 58 mm
Unit Size:
Pack of 10
Pack of 20
Product Code. 11303624
Brand: Pyrex 1200/12D
Suitable for the determination of nitrogen
  • Withstands high temperatures
  • Resistant to chemical corrosion
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner Neck 30 mm
Diameter (Metric) Outer Neck 34 mm
Capacity (Metric) 500 mL
Material Borosilicate Glass
Height (Metric) 325 mm
Neck Style Plain
Diameter (Metric) Outer 101 mm