Learn More
Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask
Product Code. p-8002554
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
125 mL
150 mL
200 mL
250 mL
300 mL
500 mL
1000 mL
2000 mL
3000 mL
5 L
Height (Metric):
52 mm
60 mm
70 mm
85 mm
105 mm
117 mm
118 mm
131 mm
145 mm
156 mm
180 mm
220 mm
280 mm
310 mm
365 mm
Diameter (Metric) Outer:
105 mm
131 mm
166 mm
187 mm
220 mm
25 mm
30 mm
42 mm
51 mm
64 mm
74 mm
79 mm
85 mm
87 mm
Body: 69 mm
Unit Size:
Each
Pack of 10
Pack of 2
Product Code. 10348669
Brand: Pyrex 1130/12D
Used for a wide range of laboratory experiments. Pyrex™ Borosilicate Glass Narrow Neck Erlenmeyer Flask is DIN ISO 1773 compliant.
  • Complies with ISO 1773 and DIN 12353
  • Erlenmeyer pattern
  • Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 1773, DIN 12353
Capacity (Metric) 200 mL
Material Borosilicate Glass
Graduations Yes
Height (Metric) 131 mm
Shape Conical
Writing Surface Yes
Graduated Yes
Diameter (Metric) Outer 79 mm
Closure Size 33