Learn More
Pyrex™ Baffled Borosilicate Glass Flask with Membrane Screw Cap
Product Code. 15507680
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Diameter (Metric) Outer:
82 mm
101 mm
129 mm
166 mm
Unit Size:
Pack of 2
Pack of 20
Pack of 5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Diameter (Metric) Outer unitSize
15507680 500 mL 101 mm Pack of 20
15272816 1000 mL 129 mm Pack of 2
15597670 250 mL 82 mm Pack of 20
15517680 2000 mL 166 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15507680 Supplier Pyrex Supplier No. 1142/10
Only null left
Add to basket
Add to basket
Ideal for aerobic microbiology and cell culture
  • Flasks complete with blue polypropylene GL45 screwcap with a gas-permeable PTFE membrane that permits gaseous exchange when contents remain sterile
  • Manufactured from Pyrex borosilicate glass or maximum chemical and thermal resistance
  • Suitable for repeated autoclave cycles at 121°C
  • Can be autoclaved with membrane screwcap fastened
  • Four equidistant baffles provide consistent agitation when used with compatible orbital shakers
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Blue
Closure Material Polypropylene
Capacity (Metric) 500 mL
Material Borosilicate Glass
For Use With (Application) Aerobic microbiology and cell culture
Thread Size GL 45
Autoclavable Autoclavable
Bottom Shape Conical
Closure Type Membrane Screw Cap
Diameter (Metric) Outer 101 mm