Learn More
Pyrex™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Product Code. p-8002539
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
Standard Taper Joint:
19/26
24/29
29/32
Unit Size:
Pack of 10
Pack of 20
Product Code. 12622062
Brand: Pyrex 1132/15
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
  • Complies to ISO 4797
  • Conical flasks with highest quality Quickfit™ ground glass socket
  • Borosilicate glass for corrosion free performance and high resistance to thermal shock
  • Erlenmeyer flasks with high quality Quickfit™ ground glass socket
  • Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance DIN 12387
Capacity (Metric) 250 mL
Material Borosilicate Glass
Graduations Yes
Height (Metric) 130 mm
Shape Conical
Standard Taper Joint 29/32
Writing Surface Yes
Graduated Yes
Diameter (Metric) Outer 85 mm